Anti-MAPT Rabbit Polyclonal Antibody

Lieferant: Abcam
AB4841-50
ABCAAB4841-50EA 810 EUR
ABCAAB4841-50
Anti-MAPT Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MAPT (microtubule-associated protein tau)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR