Anti-8-Hydroxyguanosine Mouse Monoclonal Antibody [clone: N45.1]

Lieferant: Abcam
AB48508-50
ABCAAB48508-50EA 977 EUR
ABCAAB48508-50
Anti-8-Hydroxyguanosine Mouse Monoclonal Antibody [clone: N45.1]
Antikörper
Type: Primary
Antigen: 8-Hydroxyguanosine (8-hydroxyguanosine)
Clonality: monoclonal
Clone: N45.1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR