Anti-CTDSP2 Mouse monoclonal antibody unconjugated [clone: 2230C1a]

Lieferant: Abcam
AB50621-100
ABCAAB50621-100EA 657 EUR
ABCAAB50621-100
Anti-CTDSP2 Mouse monoclonal antibody unconjugated [clone: 2230C1a]
Antikörper
Bitte beachten

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.



Type: Primary
Antigen: CTDSP2 (CTD (carboxy-terminal domain, RNA polymerase II, polypeptide A) small phosphatase 2)
Clonality: monoclonal
Clone: 2230C1a
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR