Anti-LHX4 Mouse Monoclonal Antibody [clone: LHX4E11E10]

Lieferant: Abcam
AB50790-100
ABCAAB50790-100EA 638 EUR
ABCAAB50790-100
Anti-LHX4 Mouse Monoclonal Antibody [clone: LHX4E11E10]
Antikörper
Bitte beachten

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.



Type: Primary
Antigen: LHX4 (LIM homeobox 4)
Clonality: monoclonal
Clone: LHX4E11E10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR