Anti-Ocular development associated gene Mouse Monoclonal Antibody [clone: GATA9A1D3]

Lieferant: Abcam
AB50816-100
ABCAAB50816-100EA 871 EUR
ABCAAB50816-100
Anti-Ocular development associated gene Mouse Monoclonal Antibody [clone: GATA9A1D3]
Antikörper
Type: Primary
Antigen: Ocular development associated gene
Clonality: monoclonal
Clone: GATA9A1D3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR