Anti-MMP14 Rabbit Monoclonal Antibody [clone: EP1264Y]

Lieferant: Abcam
AB51074-100
ABCAAB51074-100EA 1060 EUR
ABCAAB51074-100
Anti-MMP14 Rabbit Monoclonal Antibody [clone: EP1264Y]
Antikörper
Type: Primary
Antigen: MMP14 (matrix metallopeptidase 14 (membrane-inserted))
Clonality: monoclonal
Clone: EP1264Y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR