Anti-14-3-3 zeta Rabbit Polyclonal Antibody

Lieferant: Abcam
AB51109-100
ABCAAB51109-100EA 719 EUR
ABCAAB51109-100
Anti-14-3-3 zeta Rabbit Polyclonal Antibody
Antikörper
Bitte beachten

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.



Type: Primary
Antigen: YWHAZ (tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, zeta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR