Anti-VEGFA Rabbit Monoclonal Antibody [clone: EP1176Y]

Lieferant: Abcam
AB52917-100
ABCAAB52917-100EA 1140 EUR
ABCAAB52917-100
Anti-VEGFA Rabbit Monoclonal Antibody [clone: EP1176Y]
Antikörper
Type: Primary
Antigen: VEGFA (vascular endothelial growth factor A)
Clonality: monoclonal
Clone: EP1176Y
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR