Anti-TLR9 Goat Polyclonal Antibody

Lieferant: Abcam
AB53396-100
ABCAAB53396-100EA 943 EUR
ABCAAB53396-100
Anti-TLR9 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TLR9 (toll-like receptor 9)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR