Anti-Polyethylene glycol Rabbit Monoclonal Antibody (Biotin) [clone: PEG-B-47b]

Lieferant: Abcam
AB53449-100
ABCAAB53449-100EA 1030 EUR
ABCAAB53449-100
Anti-Polyethylene glycol Rabbit Monoclonal Antibody (Biotin) [clone: PEG-B-47b]
Antikörper
Type: Primary
Antigen: Polyethylene glycol
Clonality: monoclonal
Clone: PEG-B-47b
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR