Anti-Mycoplasma pneumoniae Rabbit Polyclonal Antibody
Lieferant: Abcam
AB53600-500
ABCAAB53600-500EA 663 EUR
ABCAAB53600-500
Anti-Mycoplasma pneumoniae Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal to Mycoplasma pneumoniae.
Bitte beachten All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Mycoplasma pneumoniae
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen