Anti-CYP3A5 Mouse Monoclonal Antibody [clone: F18 P3 B6]

Lieferant: Abcam
AB56076-100
ABCAAB56076-100EA 939 EUR
ABCAAB56076-100
Anti-CYP3A5 Mouse Monoclonal Antibody [clone: F18 P3 B6]
Antikörper
Type: Primary
Antigen: CYP3A5 (cytochrome P450, family 3, subfamily A, polypeptide 5)
Clonality: monoclonal
Clone: F18 P3 B6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR