Anti-RAD51 Rabbit Polyclonal Antibody

Lieferant: Abcam
AB61111-100
ABCAAB61111-100EA 918 EUR
ABCAAB61111-100
Anti-RAD51 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RAD51 (RAD51 recombinase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR