Anti-8-Hydroxyguanosine Mouse Monoclonal Antibody [clone: 15A3]

Lieferant: Abcam
AB62623-100
ABCAAB62623-100EA 982 EUR
ABCAAB62623-100
Anti-8-Hydroxyguanosine Mouse Monoclonal Antibody [clone: 15A3]
Antikörper
Type: Primary
Antigen: 8-Hydroxyguanosine (8-hydroxyguanosine)
Clonality: monoclonal
Clone: 15A3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR