Anti-Agmatine Chicken Polyclonal Antibody

Lieferant: Abcam
AB62667-100
ABCAAB62667-100EA 871 EUR
ABCAAB62667-100
Anti-Agmatine Chicken Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Agmatine
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR