Anti-HLA DR Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: MEM-267]

Lieferant: Abcam
AB64676-100
ABCAAB64676-100EA 913 EUR
ABCAAB64676-100
Anti-HLA DR Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: MEM-267]
Antikörper
Type: Primary
Antigen: HLA DR
Clonality: monoclonal
Clone: MEM-267
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR