Anti-TNFRSF10A Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: DR-4-02]

Lieferant: Abcam
AB65810-50
ABCAAB65810-50EA 912 EUR
ABCAAB65810-50
Anti-TNFRSF10A Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: DR-4-02]
Antikörper
Type: Primary
Antigen: TNFRSF10A (tumor necrosis factor receptor superfamily, member 10a)
Clonality: monoclonal
Clone: DR-4-02
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR