Anti-LAT Rabbit Monoclonal Antibody [clone: EP983Y]

Lieferant: Abcam
AB68139-100
ABCAAB68139-100EA 855 EUR
ABCAAB68139-100
Anti-LAT Rabbit Monoclonal Antibody [clone: EP983Y]
Antikörper
Type: Primary
Antigen: LAT (linker for activation of T cells)
Clonality: monoclonal
Clone: EP983Y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR