Anti-IgG Donkey Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))

Lieferant: Abcam
AB6817-1
ABCAAB6817-1EA 350 EUR
ABCAAB6817-1
Anti-IgG Donkey Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Antikörper
Donkey Anti-Mouse IgG H&L (TRITC).

Bitte beachten All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: TRITC (Tetramethylrhodamine Isothiocyanate)
Epitope:
Host: Donkey
Isotype:
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR