Anti-IgG Sheep Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))

Lieferant: Abcam
AB6867-1
ABCAAB6867-1EA 350 EUR
ABCAAB6867-1
Anti-IgG Sheep Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Antikörper
Sheep Anti-Human IgG H&L (TRITC).

Bitte beachten All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: TRITC (Tetramethylrhodamine Isothiocyanate)
Epitope:
Host: Sheep
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR