Anti-IgG Rabbit Polyclonal Antibody (Texas Red®)

Lieferant: Abcam
AB6919-1
ABCAAB6919-1EA 396 EUR
ABCAAB6919-1
Anti-IgG Rabbit Polyclonal Antibody (Texas Red®)
Antikörper
Type: Secondary
Antigen: IgG (Horse) (horse IgG)
Clonality: polyclonal
Clone:
Conjugation: Texas Red®
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR