Anti-IgG Goat Polyclonal Antibody (Cy5.5®)

Lieferant: Abcam
AB6942-500
ABCAAB6942-500EA 471 EUR
ABCAAB6942-500
Anti-IgG Goat Polyclonal Antibody (Cy5.5®)
Antikörper
Type: Secondary
Antigen: IgG (Rabbit) (rabbit IgG)
Clonality: polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Goat
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR