Anti-Nitro tyrosine Mouse Monoclonal Antibody [clone: HM.11]

Lieferant: Abcam
AB7048-50
ABCAAB7048-50EA 732 EUR
ABCAAB7048-50
Anti-Nitro tyrosine Mouse Monoclonal Antibody [clone: HM.11]
Antikörper
Bitte beachten

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.



Type: Primary
Antigen: Nitro tyrosine
Clonality: monoclonal
Clone: HM.11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR