Anti-GABA Transporter 1 / GAT 1 Rabbit polyclonal antibody unconjugated
Lieferant: Abcam
AB72448-100
ABCAAB72448-100EA 943 EUR
ABCAAB72448-100
Anti-GABA Transporter 1 / GAT 1 Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: GABA Transporter 1 / GAT 1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat