Anti-V5 Tag Rabbit Polyclonal Antibody (SureLight® Allophycocyanin)

Lieferant: Abcam
AB72560-100
ABCAAB72560-100EA 930 EUR
ABCAAB72560-100
Anti-V5 Tag Rabbit Polyclonal Antibody (SureLight® Allophycocyanin)
Antikörper
Type: Primary
Antigen: V5 Tag (V5 tag)
Clonality: polyclonal
Clone:
Conjugation: SureLight® Allophycocyanin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR