Anti-T7 Tag Rabbit Polyclonal Antibody (SureLight® Allophycocyanin)

Lieferant: Abcam
AB72565-100
ABCAAB72565-100EA 918 EUR
ABCAAB72565-100
Anti-T7 Tag Rabbit Polyclonal Antibody (SureLight® Allophycocyanin)
Antikörper
Type: Primary
Antigen: T7 Tag (T7 tag)
Clonality: polyclonal
Clone:
Conjugation: SureLight® Allophycocyanin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR