Anti-GST Goat Polyclonal Antibody SureLight® Allophycocyanin

Lieferant: Abcam
AB72568-100
ABCAAB72568-100EA 912 EUR
ABCAAB72568-100
Anti-GST Goat Polyclonal Antibody SureLight® Allophycocyanin
Antikörper
Type: Primary
Antigen: GST
Clonality: polyclonal
Clone:
Conjugation: SureLight® Allophycocyanin
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR