Anti-DDDDK tag Mouse monoclonal antibody SureLight® Allophycocyanin [clone: M2]

Lieferant: Abcam
AB72569-100
ABCAAB72569-100EA 918 EUR
ABCAAB72569-100
Anti-DDDDK tag Mouse monoclonal antibody SureLight® Allophycocyanin [clone: M2]
Antikörper
Type: Primary
Antigen: DDDDK tag (DDDDK tag)
Clonality: monoclonal
Clone: M2
Conjugation: SureLight® Allophycocyanin
Epitope:
Host: Mouse
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR