Anti-PDCD5 Rabbit polyclonal antibody

Lieferant: Abcam
AB75430-100
ABCAAB75430-100EA 0 EUR
ABCAAB75430-100
Anti-PDCD5 Rabbit polyclonal antibody
Antikörper
Type: Primary
Antigen: PDCD5 (programmed cell death 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR