Anti-Nicotinic Acetylcholine Receptor beta Rabbit Monoclonal Antibody [clone: EP2067Y]

Lieferant: Abcam
AB76159-100
ABCAAB76159-100EA 862 EUR
ABCAAB76159-100
Anti-Nicotinic Acetylcholine Receptor beta Rabbit Monoclonal Antibody [clone: EP2067Y]
Antikörper
Bitte beachten

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.



Type: Primary
Antigen: Nicotinic Acetylcholine Receptor beta
Clonality: monoclonal
Clone: EP2067Y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR