Anti-Cyanine Sheep Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Abcam
AB7627-100
ABCAAB7627-100EA 638 EUR
ABCAAB7627-100
Anti-Cyanine Sheep Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
HRP Sheep polyclonal to Cyanine.

Bitte beachten All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Cyanine
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Sheep
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR