Anti-HLA G Mouse Monoclonal Antibody [clone: 5A6G7]

Lieferant: Abcam
AB76869-100
ABCAAB76869-100EA 748 EUR
ABCAAB76869-100
Anti-HLA G Mouse Monoclonal Antibody [clone: 5A6G7]
Antikörper
Type: Primary
Antigen: HLA G
Clonality: monoclonal
Clone: 5A6G7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR