Anti-HLA DR + HLA DP Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: MEM-136]

Lieferant: Abcam
AB77134-100
ABCAAB77134-100EA 912 EUR
ABCAAB77134-100
Anti-HLA DR + HLA DP Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: MEM-136]
Antikörper
Type: Primary
Antigen: HLA DR + HLA DP
Clonality: monoclonal
Clone: MEM-136
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR