Anti-Dopamine Transporter Rabbit Polyclonal Antibody

Lieferant: Abcam
AB78068-50
ABCAAB78068-50EA 918 EUR
ABCAAB78068-50
Anti-Dopamine Transporter Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Dopamine Transporter
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR