Anti-Activin A Receptor Type IB/ALK-4 Mouse Monoclonal Antibody [clone: MM0054-11A7]

Lieferant: Abcam
AB78415-100
ABCAAB78415-100EA 657 EUR
ABCAAB78415-100
Anti-Activin A Receptor Type IB/ALK-4 Mouse Monoclonal Antibody [clone: MM0054-11A7]
Antikörper
Mouse monoclonal [MM0054-11A7] to Activin A Receptor Type IB/ALK-4.

Bitte beachten All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Activin A Receptor Type IB/ALK-4
Clonality: Monoclonal
Clone: MM0054-11A7
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR