Anti-L1CAM Mouse Monoclonal Antibody [clone: UJ127]

Lieferant: Abcam
AB80832-100
ABCAAB80832-100EA 800 EUR
ABCAAB80832-100
Anti-L1CAM Mouse Monoclonal Antibody [clone: UJ127]
Antikörper
Mouse monoclonal [UJ127] to L1CAM - BSA and Azide free.

Bitte beachten All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: L1CAM
Clonality: Monoclonal
Clone: UJ127
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR