Anti-DDDDK tag (Binds to FLAG® tag sequence) Mouse Monoclonal Antibody [clone: M2] AP (Alkaline Phosphatase)

Lieferant: Abcam
AB81655-100
ABCAAB81655-100EA 688 EUR
ABCAAB81655-100
Anti-DDDDK tag (Binds to FLAG® tag sequence) Mouse Monoclonal Antibody [clone: M2] AP (Alkaline Phosphatase)
Antikörper
Alkaline Phosphatase Mouse monoclonal [M2] to DDDDK tag (Binds to FLAG® tag sequence).

Bitte beachten All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: DDDDK tag (Binds to FLAG® tag sequence)
Clonality: Monoclonal
Clone: M2
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR