Anti-Histone H3 (asymmetric di methyl R17) Rabbit Polyclonal Antibody

Lieferant: Abcam
AB8284-50
ABCAAB8284-50EA 488 EUR
ABCAAB8284-50
Anti-Histone H3 (asymmetric di methyl R17) Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal to Histone H3 (asymmetric di methyl R17).

Bitte beachten All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Histone H3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR