Anti-IL-15 Rabbit Polyclonal Antibody (Biotinylated)

Lieferant: Abcam
AB83157-50
ABCAAB83157-50EA 663 EUR
ABCAAB83157-50
Anti-IL-15 Rabbit Polyclonal Antibody (Biotinylated)
Antikörper
Biotin Rabbit polyclonal to IL-15.

Bitte beachten All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: IL-15
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Recombinant full length protein
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR