Anti-CALCRL Rabbit polyclonal antibody

Lieferant: Abcam
AB84467-100
ABCAAB84467-100EA 0 EUR
ABCAAB84467-100
Anti-CALCRL Rabbit polyclonal antibody
Antikörper
Type: Primary
Antigen: CALCRL (calcitonin receptor-like)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR