Anti-Monocarboxylic acid transporter 1 Rabbit Polyclonal Antibody

Lieferant: Abcam
AB85021-100
ABCAAB85021-100EA 953 EUR
ABCAAB85021-100
Anti-Monocarboxylic acid transporter 1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Monocarboxylic acid transporter 1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR