Anti-CD8A Rabbit polyclonal antibody unconjugated

Lieferant: Abcam
AB85792-100
ABCAAB85792-100EA 826 EUR
ABCAAB85792-100
Anti-CD8A Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: CD8A (CD8a molecule)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR