Anti-NuMA Rabbit Polyclonal Antibody

Lieferant: Abcam
AB86129-100
ABCAAB86129-100EA 954 EUR
ABCAAB86129-100
Anti-NuMA Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NuMA
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR