Anti-Histone H4 (di methyl K20) Rabbit Polyclonal Antibody

Lieferant: Abcam
AB9052-100
ABCAAB9052-100EA 532 EUR
ABCAAB9052-100
Anti-Histone H4 (di methyl K20) Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal to Histone H4 (di methyl K20).

Bitte beachten All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Histone H4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Bovine;Human;S. pombe;Toxoplasma gondii;Drosophila melanogaster
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR