Anti-HLA DR Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: GRB-1]

Lieferant: Abcam
AB91334-100
ABCAAB91334-100EA 916 EUR
ABCAAB91334-100
Anti-HLA DR Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: GRB-1]
Antikörper
Type: Primary
Antigen: HLA DR
Clonality: monoclonal
Clone: GRB-1
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR