Anti-Prostaglandin E Receptor EP4 Rabbit Polyclonal Antibody (SureLight® Allophycocyanin)

Lieferant: Abcam
AB92763-50
ABCAAB92763-50EA 912 EUR
ABCAAB92763-50
Anti-Prostaglandin E Receptor EP4 Rabbit Polyclonal Antibody (SureLight® Allophycocyanin)
Antikörper
Type: Primary
Antigen: Prostaglandin E Receptor EP4
Clonality: polyclonal
Clone:
Conjugation: SureLight® Allophycocyanin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR