Anti-MAPT Rabbit Monoclonal Antibody [clone: SP70]

Lieferant: Abcam
AB93726-100
ABCAAB93726-100EA 980 EUR
ABCAAB93726-100
Anti-MAPT Rabbit Monoclonal Antibody [clone: SP70]
Antikörper
Type: Primary
Antigen: MAPT (microtubule-associated protein tau)
Clonality: monoclonal
Clone: SP70
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR