Anti-CCR5 Armenian Hamster Monoclonal Antibody (PE (Phycoerythrin)) [clone: HM-CCR5(7A4)]

Lieferant: Abcam
AB95662-50
ABCAAB95662-50EA 344 EUR
ABCAAB95662-50
Anti-CCR5 Armenian Hamster Monoclonal Antibody (PE (Phycoerythrin)) [clone: HM-CCR5(7A4)]
Antikörper
Bitte beachten

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.



Type: Primary
Antigen: CCR5 (chemokine (C-C motif) receptor 5 (gene/pseudogene))
Clonality: monoclonal
Clone: HM-CCR5(7A4)
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR