Anti-COUP TF1 Rabbit Polyclonal Antibody

Lieferant: Abcam
AB96846-50
ABCAAB96846-50EA 939 EUR
ABCAAB96846-50
Anti-COUP TF1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: COUP TF1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR