Anti-IgG2b Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Abcam
AB98703-500
ABCAAB98703-500EA 430 EUR
ABCAAB98703-500
Anti-IgG2b Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Type: Secondary
Antigen: IgG2b (Mouse) (mouse IgG2b)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR